Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAPK6 IgG Polyclonal Antibody

Supplier: Boster
PB9666
76173-796EA 698.31 USD InStock
76173-796
Anti-MAPK6 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Mitogen-activated protein kinase 6(MAPK6) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Mitogen-activated protein kinase 6 is an enzyme that in humans is encoded by the MAPK6 gene. It is mapped to 15q21. The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblastsupon treatment with serum or phorbol esters.

Type: Primary
Antigen: MAPK6
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty