Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HDAC7 IgG Polyclonal Antibody

Supplier: Boster
PB9629
76174-114EA 622.93 USD InStock
76174-114
Anti-HDAC7 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Histone deacetylase 7(HDAC7) detection. Tested with WB in Human;Rat.

Histone deacetylase 7 is an enzyme that in humans is encoded by the HDAC7 gene. It is mapped to 12q13.1. Histones play a critical role in transcriptional regulation, cell cycle progression, and developmental events. Histone acetylation / deacetylation alters chromosome structure and affects transcription factor access to DNA. The protein encoded by this gene has sequence homology to members of the histone deacetylase family. This gene is orthologous to mouse HDAC7 gene whose protein promotes repression mediated via the transcriptional corepressor SMRT. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na₂HPO₄, 0.05 mg NaN₃.

Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: HDAC7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty