Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HHEX IgG Polyclonal Antibody

Supplier: Boster
PB9631
76174-118EA 622.93 USD InStock
76174-118
Anti-HHEX IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Hematopoietically-expressed homeobox protein Hhex(HHEX) detection. Tested with WB in Human;Mouse; Rat.

Hematopoietically-expressed homeobox protein HHEX is a protein that in humans is encoded by the HHEX gene. Homeobox genes are members of a family of transcription factors that regulate tissue development in many different organisms. Hromas et al. (1993) set out to identify homeobox genes that might play a role in hematopoiesis. And using somatic cell hybrid analysis, they mapped the HHEX gene to chromosome 10, where the HOX11 gene is located. Homeobox genes are involved in neoplastic transformation of both epithelial and hemopoietic tissues. The divergent homeobox gene HEX is expressed in the anterior visceral endoderm during early mouse development and in some adult tissues of endodermal origin, including liver and thyroid. D'Elia et al.’s findings suggested that regulation of HEX entry in the nucleus of thyrocytes may represent a critical step during human thyroid tumorigenesis.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Each vial contains 5 mg BSA, 0.9 mg NaCl, 0.2 mg Na₂HPO₄, 0.05 mg NaN₃.

Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.

Type: Primary
Antigen: HHEX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human; Mouse; Rat

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty