Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AEBP2 IgG Polyclonal Antibody

Supplier: Boster
PB9977
76174-416EA 698.31 USD InStock
76174-416
Anti-AEBP2 IgG Polyclonal Antibody
Antibodies
Rabbit IgG polyclonal antibody for Zinc finger protein AEBP2(AEBP2) detection. Tested with WB in Human.

1. Imhof, Axel; Kim, Hana; Bakshi, Arundhati; Kim, Joomyeong (2015). "Retrotransposon-Derived Promoter of Mammalian Aebp2". PLOS ONE 10 (4): e0126966.
2. Kim H, Kang K, Kim J (2009). "AEBP2 as a potential targeting protein for Polycomb Repression Complex PRC2". Nucleic Acids Res. 37(9): 2940–50.

Add 0.2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: AEBP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty