Would you like to log in again?
West Nile Virus Recombinant WNV NS3 Protease (from E. coli)
Supplier: AnaSpec
AS-72081-5 AS-72081-100
103010-402EA 818.73 USD InStock
103010-402 103010-404
West Nile Virus Recombinant WNV NS3 Protease (from E. coli)
Proteins and Peptides
West Nile virus (WNV) is a member of the flavivirus genus, which contains many significant human pathogens including Dengue virus (Den), Japanese Encephalitits virus (JE), and Yellow Fever virus (YF). WNV is a small, enveloped virus with a single stranded, positive sense 11kb RNA genome, which encodes a polyprotein precursor. This polyprotein must be cleaved co- and post-translationally to produce ten functional proteins: three structural (C, prM and E) and 7 nonstructural (NS1, NS2A, NS2B, NS3, NS4A,NS4B and NS5). WNV NS3 protease is absolutely essential (along with viral-encoded cofactor NS2B) for post-translational cleavage and viral replication. As a result, this protease is a potential therapeutic target.
His-tagged recombinant WNV NS3 protease (residues 1 to 184) was expressed as a fusion protein with the cofactor NS2b (residues 49 to 96) and a linker sequence (GGGGSGGGG) in E. coli. The apparent Mr on SDS-PAGE is 32-kDa. Its activity can be measured by FRET peptides
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty