Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]

Supplier: BioGems
04111-25-50
75841-614EA 303.61 USD InStock
75841-614
Anti-CD2 Mouse Monoclonal Antibody [clone: RPA-2.10]
Antibodies
The RPA-2.10 monoclonal antibody specifically binds to human CD2, a 50 kDA type I transmembrane glycoprotein. CD2 is expressed on NK cells, thymocytes, T lymphocytes, and B cell subsets. CD2 is involved in cell-cell adhesion, T cell activation, and T cell signaling. Its ligand is CD58 and it is reported to bind to CD48, CD59, and CD15. The RPA-2.10 antibody is reported to be cross-reactive with pigs and non-human primates..

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty