Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PD-1 Armenian Hamster Monoclonal Antibody [clone: J43.1]

Supplier: BioGems
31812-20-100
75843-268EA 172.49 USD InStock
75843-268
Anti-PD-1 Armenian Hamster Monoclonal Antibody [clone: J43.1]
Antibodies
The J43.1 monoclonal antibody specifically reacts with mouse CD279, also known as PD-1 (programmed death-1), a 50-55 kDa glycoprotein of the Ig superfamily. The PD-1 ligands, PD-L1 (B7-H1) and PD-L2 (B7-H2) are members of the B7 family. Pd-1 contains an Immunoreceptor Tyrosine-based Inhibitory Motif (ITIM) and influences the peripheral tolerances and autoimmune diseases in mice. PD-1 is transiently expressed on CD4/CD8 thymocytes, it is upregulated in apoptotic cells, and it is expressed by activated myeloid and T and B cells. The binding of PD-1 to its ligands is blocked by the J43 antibody, which also enhances contact hypersensitivity and exacerbates acute Graft-versus-host disease, Experimental autoimmune encephalomyelitis and NOD diabetes. PD-1 seems to downregulate the immune response, as the development of splenomegaly and breakdown of peripheral tolerance in PD-1 deficient mice suggests.

Type: Primary
Antigen: PD-1
Clonality: Monoclonal
Clone: J43.1
Conjugation:
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty