Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: A20]

Supplier: BioGems
07522-77-25
75844-150EA 70.27 USD InStock
75844-150
Anti-CD45.1 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: A20]
Antibodies
The A20 monoclonal antibody reacts with the leukocytes of the CD45.1-expressing mouse strains (DA, SJL/J, RIII, and STS/A). It does not cross-react with cells that express CD45.2. The CD45 molecule is a member of the Protein Tyrosine Phosphatase (PTP) family, because its intracellular region contains two PTP domains. The extracellular region’s variability is caused by different levels of glycosylation, and the splicing of the 4, 5, and 6 exons. The isoforms found in the mouse strains depend on the activation state, maturation stage and cell type, and are very important in B and T lymphocytes antigen receptor signal transduction. The A20 antibody inhibits some of the B lymphocytes responses, from CD45.1-expressing mice, to lipopolysaccharides and antigens.

Type: Primary
Antigen: CD45.1
Clonality: Monoclonal
Clone: A20
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty