Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Foxp3 (3G3) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3G3]

Supplier: BioGems
83412-60-100
75842-602EA 421.63 USD InStock
75842-602
Anti-Foxp3 (3G3) Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 3G3]
Antibodies
The 3G3 monoclonal antibody specifically reacts with the mouse 50-55 kDa Foxp3 protein (JM2, IPEX), a member of the forkhead family of transcription factors. Foxp3 is expressed by the Treg lymphocytes, whose development and function are influenced by the forkhead protein. Ectopic expression of Foxp3 in T lymphocytes inhibits their activity and cytokine expression.Mutations of Foxp3 result in the “scurfy” mice phenotype.

Type: Primary
Antigen: Foxp3 (3G3)
Clonality: Monoclonal
Clone: 3G3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty