Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-TNFA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP6-XT22]

Supplier: BioGems
84422-80-50
75841-398EA 354.64 USD InStock
75841-398
Anti-TNFA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MP6-XT22]
Antibodies
The MP6-XT22 monoclonal antibody specifically reacts with mouse tumor necrosis factor alpha (TNF-alpha), a 156 amino pro-inflammatory cytokine. It is secreted by various cells including adipocytes, activated monocytes, macrophages, B cells, T cells and fibroblasts. NF-α is cytotoxic to a wide variety of tumor cells and is an essential factor in mediating the immune response against bacterial infections. TNF-α also plays a role in the induction of septic shock, auto immune diseases, rheumatoid arthritis, inflammation, and diabetes. The MP6-XT22 is reported to neutralize the bioactivity of natural or recombinant TNF-alpha and is useful for intracellular immunofluorescent staining.

Type: Primary
Antigen: TNFA
Clonality: Monoclonal
Clone: MP6-XT22
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG1, kappa
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty