Would you like to log in again?
Clip-On Welders Adapter withCover Lenses, MSA
Supplier: MSA
454-472859
300081-070EA 421.31 USD InStock
300081-070
Clip-On Welders Adapter withCover Lenses, MSA
Welding Supplies
The Clip-on Welder's Adapter for Ultravue and Ultra-Twin® facepieces is supplied with a cover lens
  • Clip-on adapter attaches to facepieces by molded in clips
  • Integral adapter attaches by removing facepiece lens and replacing with adapter
MSA Welder's Adapters are molded by polycarbonate plastic, which has high resistance to impact, heat, and welding splatter.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty