Would you like to log in again?
Dial Caliper and Micrometer Set, L.S. STARRETT, ORS Nasco
Supplier: ORS Nasco
681-65122
300042-811EA 611.67 USD InStock
300042-811
Dial Caliper and Micrometer Set, L.S. STARRETT, ORS Nasco
Tool Sets
Meets or Exceeds Accuracy and Performance Requirements of National and International Standards and are traceable to the National Institute of Standards and Technology.
Delivery information: Set includes 120A-6 Dial Caliper, C604R-6 Spring Tempered Precision Rule, T436.1XRL-1 Outside Micrometer with Carbide Faces, and case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty