Would you like to log in again?
Flexi-Filter® Pads for Advantage® Respirators, MSA
Supplier: MSA
Flexi-Filter®
454-818342 454-818343
300009-025PK 19.37 USD InStock
300009-025 300009-026
Flexi-Filter® Pads for Advantage® Respirators, MSA
Respirators Reusable Respirators Respirator Filters
Swept-back design provides improved vision and comfort.
  • Low-profile, flexible pad design
  • Fits well under a welding hood and with other personal protective equipment
Certifications: NIOSH Approved to 42 CFR Part 84.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty