Would you like to log in again?
Pre-cut PEEK Tubing, Thermo Scientific
Supplier: Thermo Scientific
37005
MSPP-37005PK 72.31 USD InStock
MSPP-37005
Pre-cut PEEK Tubing, Thermo Scientific
Tubing Chromatography Tubing
This tubing is designed to work in most HPLC applications, particularly reversed phase with aqueous mobile phases.
  • Resistant to most organic solvents
  • Available in a range of internal diameters
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty