Would you like to log in again?
Azlon® Wash Bottle, Sloped Shoulder, Printed, Dynalon
Supplier: Dynalon
Azlon®
506915-0001
80700-216CS 58.91 USD InStock
80700-216
Azlon® Wash Bottle, Sloped Shoulder, Printed, Dynalon
Bottles Wash Bottles
Sloped shoulder, 'Water' printed wash bottle and color-coded polypropylene leak-proof closure.
  • Closure features an acute angled 'swan neck' dispensing spout
  • Maximum temperature of LDPE is 80 °C
Stylish sloped shoulder LDPE printed wash bottles are supplied with a color-coded polypropylene swivel/leak-proof closure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty