Would you like to log in again?
Human Peripheral Blood CD3+ T Cells (Pan T Cells), iXCells Biotechnologies
Supplier: iXCells Biotechnologies
10HU-009-20M 10HU-009-40M
103521-814EA 1803.03 USD InStock
103521-814 103521-812
Human Peripheral Blood CD3+ T Cells (Pan T Cells), iXCells Biotechnologies
Cells
Human Peripheral Blood CD3+ T Cells (Pan T Cells); Frozen
  • High-quality Human Peripheral Blood CD3+ T Cells (Pan T Cells)
  • High purity
  • IRB-approved
CD3+ Pan T Cells are isolated from human peripheral blood mononuclear cells (PBMCs) using negative immunomagnetic selection and are untouched by the separation process. > 86% of the cells are CD3 positive.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty