Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Human IPSC Growth Medium, iXCells Biotechnologies
Supplier: iXCells Biotechnologies
MD-0018-500ML
103521-474EA 303.88 USD InStock
103521-474
Human IPSC Growth Medium, iXCells Biotechnologies
Cell Culture Media
Human IPSC Growth Medium designed for human induced pluripotent stem cells (hiPSCs) culture.
  • Optimized for human pluripotent stem cells (hPSCs) culture
  • Medium is fully supplemented and sterile
  • Read-to-use
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty