Would you like to log in again?
Senescence Kit, OZ Biosciences
Supplier: OZ Biosciences
GXS0003
103258-160EA 242.08 USD InStock
103258-160
Senescence Kit, OZ Biosciences
Assays Cellular Assays
Senescence Kit is ideal when cells in culture showed abnormalities typical of the Hayflick limit, after a long period of normal growth
  • Simple and Rapid
  • Ready-to-use
  • Economical
Senescence goes with decreasing of proliferation potential, telomere shortening and impairment of cell function (i.e decrease of differentiation potential of stem cells). During senescence in mammalian cells, an endogenous lysosomal b-galactosidase is over-expressed and is accumulated within the aging cells. The presence of this b-Gal activity is a marker of aging cell population in vitro. 
The cells vary in size and shape, the cytoplasm begins to be granular with many cell inclusions and debris formed in the medium.
Delivery information: Shipped with a gel pack
Packaging: Store at +4°C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty