Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (FITC)

Supplier: OZ Biosciences
IGG0100
103258-162EA 124.05 USD InStock
103258-162
Anti-IgG Goat Polyclonal Antibody (FITC)
Antibodies
OZ Biosciences Immunoglobulin (IgG) from goat serum has been labelled with the fluorescein isothiocyanate dye to produce a green-emitting antibody with a high fluorophore-to-protein ratio. The resulting antibody can be used as a positive control for intracellular antibody delivery with OZ Biosciences’ Ab-DeliverIN reagent (cat. #AI21000).

  • Absorption Maximum: 492 nm
  • Emission maximum: 520nm
  • Extinction coefficient: 75000 M-1cm-1
  • Quantum yield: 0.79

Delivery information: Shipped at room temperature

Packaging: Store at -20°C

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty