Would you like to log in again?
MTT Cell Proliferation assay kit , OZ Biosciences
Supplier: OZ Biosciences
MT01000
103258-310EA 264.23 USD InStock
103258-310
MTT Cell Proliferation assay kit , OZ Biosciences
Assays Cellular Assays
OZ Biosciences MTT Cell Proliferation Assay Kit is designed for spectrophotometric quantification of cell growth, viability and proliferation and can be used as a direct indicator of cytotoxicity (such as for screening anticancer drugs) and apoptosis
  • Easy and Ready to use
  • Accurate: The absorbance signal produced correlates with the cell number
  • Convenient packaging
  • Economical
The MTT Cell Proliferation Kit is a colorimetric assay for measuring the mitochondrial reductases activity in living cells. It is based on the cleavage of membrane-permeable yellow tetrazolium salt MTT to blue/purple formazan crystals by metabolically active cells. This MTT assay kit contains sufficient reagents to perform 1000 assays in 96-well plate format.
Delivery information: Shipped at room temperature
Packaging: Store MTT Reagent at -20°C MTT Reagent  / Solubilization solution at +4°C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty