Would you like to log in again?
ONPG assay kit, OZ Biosciences
Supplier: OZ Biosciences
GO10001
103258-322EA 293.69 USD InStock
103258-322
ONPG assay kit, OZ Biosciences
Assays Enzyme Assays
OZ Biosciences ONPG assay kit is ideal to quantify high expression level of beta-Gal
  • Convenient for all transfection assays
  • Suitable for cultured cells and tissues
  • Simple, rapid and easy-to-use
  • Economical
  • Optimized with stable buffers
All the necessary reagents are provided in this assay kit and offer a rapid, simple and sensitive method to quantify spectrophotometrically the enzyme expression level in LacZ transfected cells.
The levels of active beta-Gal expression can be quickly measured by its catalytic hydrolysis of ONPG (o-nitrophenyl-beta-D-galactopyranoside) substrate to a bright yellow product (Absorbance at 405-420nm).LacZ is one of the most frequently reporter gene used in transfection experiments. The LacZ encoded protein (beta-galactosidase, beta-Gal), is very stable, resistant to proteolytic degradation and easily assayed. 
Delivery information: Shipped with a gel pack
Packaging: Store all reagents at +4°C except Beta Gal Enzyme
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty