Would you like to log in again?
OZBlue Cell Viability kit , OZ Biosciences
Supplier: OZ Biosciences
BL00025 BL00100
103258-430EA 473.71 USD InStock
103258-430 103258-428
OZBlue Cell Viability kit , OZ Biosciences
Assays Cellular Assays
Check cell viability without killing cells
  • Non-toxic: Allows additional analysis on proliferating cells (mRNA, cytogenetic, apoptosis, immunophenotyping,...) and continuous monitoring of cultures over the time
  • High Sensitivity: More sensitive than (3H)Thymidine incorporation and Tetrazolium reduction Assay (MTT, XTT)
  • Simple: a single ready-to-use reagent. No need of washing or extraction procedures
  • Versatile: can be used on cell lines, primary cells, bacteria, fungi
  • Flexible: Colorimetric and fluorescence detection allows compativility with flow cytometry, plate readers and microscopy detection
  • Rapid & Cost effective
The OZBlue Cell Viability kit is a ready-to-use assay system based on fluorimetric/colorimetric quantification of metabolic activity in living cells. The Resazurin dye is used as an indicator of cell viability.
Delivery information: Shipped with ice pack
Packaging: Store at +4°C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty