Would you like to log in again?
Luciferase Assay kit , OZ Biosciences
Supplier: OZ Biosciences
LUC1000 LUC0100
103258-432EA 146.37 USD InStock
103258-432 103258-434
Luciferase Assay kit , OZ Biosciences
Assays Enzyme Assays
The luciferase assay kit is the most sensitive analytical tools for measuring gene expression
  • Very sensitive and quantitative assay with a wide dynamic range
  • Bioluminescence measurements are performed directly into transfected wells
  • Cell lysis buffer is optimized for Luciferase assay buffer to ensure maximal sensitivity
  • Light emission is proportional to number of luciferase enzyme molecule
Luciferase assay kit contains optimized components (cell lysis buffer, luciferase substrate and Luciferase Assay Buffer) to ensure maximal sensitivity with a consistent light output. 
Fast and easy, accurate and linear, this kit detects and quantify firefly luciferase in transfected eukaryotic cells reaching high sensitivity level. Since the light emission is constant (>15 min) when using this kit, there is no need to use an automated luminometer for substrate injection. Emitted light is recorded by a standard luminometer at 560nm.
Delivery information: Shipped with dry ice
Packaging: Store at +4°C / -20 °C
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty