Would you like to log in again?
Air Scribe® Kit, Chicago Pneumatic
Supplier: Chicago Pneumatic
Air Scribe®
147-93611
300042-971EA 722.38 USD InStock
300042-971
Air Scribe® Kit, Chicago Pneumatic
Pens Stylus and Scribers
Adjustable power for a variety of applications and materials such as metals, plastics, ceramics, and glass.
  • Convertible to a hammer or a light scaler
  • Lightweight for easy operation
  • 5.1 m/s2 vibration
  • 1/4" (NPT) inlet size
Cycle rate is 13,500 blows/min. Air consumption is 1.1 CFM.
Delivery information: Kit includes air filter assembly, air hose, anvil, use with chisels & blanks, carrying case, chisel blank, flat chisel, round nose chisel, stylus, carbide tipped.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty