Would you like to log in again?
Choice® Edge Cleanroom-Laundered, Bordered, Sealed-Edge 100% Polyester Knit Wiper, Berkshire
Supplier: Berkshire
CHOICE®
CHKE09.14
76193-156CS 644.37 USD InStock
76193-156
Choice® Edge Cleanroom-Laundered, Bordered, Sealed-Edge 100% Polyester Knit Wiper, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Smooth knit construction and consistent border with sealed edges for scratch-free wiping.
  • No-run interlock construction
  • Sealed edges for increased durability
Wipers are made from 100% polyester knit. Four-side-bordered, with sealed edges to increase durability and minimize fiber and particle release.
Certifications: Laundered in an ISO Class 4 (Class 10) Cleanroom.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty