Would you like to log in again?
Chymotrypsin Activity Assay Kit (Fluorometric), Biovision
Supplier: BioVision
AB234051-100TEST
76186-146EA 808.2 USD InStock
76186-146
Chymotrypsin Activity Assay Kit (Fluorometric), Biovision
Assays Enzyme Assays
Simple method to measure specific chymotrypsin activity in samples containing non-specific proteases
  • Simple procedure; takes ~ 1-2 hours
  • Fast and convenient
Chymotrypsin (EC 3.4.21.1) is a key serine protease involved in dietary protein digestion in mammals. It is primarily produced by the pancreas, but may be expressed in other tissues, including spleen and liver. In the pancreas, chymotrypsin is initially expressed as the inactive proenzyme chymotrypsinogen, which is cleaved by other proteases to chymotrypsin, its active form. Chymotrypsin specifically cleaves peptide bonds at the C-terminal end of bulky hydrophobic or aromatic amino acids (such as tyrosine, tryptophan or phenylalanine). BioVision’s Chymotrypsin Activity Assay kit uses a synthetic fluorogenic substrate, enabling kinetic measurement of chymotrypsin activity in cell and tissue lysates. A chymotrypsin activator cleaves chymotrypsinogen to form active chymotrypsin, which then hydrolyzes the non-fluorescent substrate to release a stable fluorophore (Ex/Em = 380/460 nm). The kit includes a selective chymotrypsin inhibitor that can be used to measure specific chymotrypsin activity in samples containing non-specific proteases and endopeptidases that may also metabolize the substrate. The assay can detect as low as 0.01 mU of Chymotrypsin
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty