Would you like to log in again?
Mitochondrial DNA Purification Kit, Biovision, Inc.
Supplier: BioVision
AB288088-25TEST
76186-206EA 355.63 USD InStock
76186-206
Mitochondrial DNA Purification Kit, Biovision, Inc.
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Yields highly purified mt
  • Simple procedure; takes less than 2 hours
  • Fast and convenient
  • The kit provides unique formulations of buffers, reagents anc columns for isolating a highly enriched mitochondrial fraction. This kit provides genomic-free DNA samples
Mitochondria are essential double-membrane bound organelles that support aerobic respiration and most of the cellular ATP production in all nucleated mammalian cells. To date, hundreds of pathogenic mitochondrial DNA mutations have been identified to result in disease phenotypes such as cancer, ataxia, and diabetes. As a result, isolation of high quality mitochondrial DNA from mammalian cells and tissues in sufficient quantities is important for many basic research and clinical applications. BioVision’s Mitochondrial DNA Isolation Kit facilitates purification of quality mitochondrial DNA from mammalian cultured cells or tissue. This kit utilizes a density separation step to isolate mitochondria from the rest of the cellular organelles, followed by an enzymatic reaction to release DNA from the mitochondria. Upon release, mitochondrial DNA is further purified from nuclear DNA contamination and adsorbed onto a silica spin-column under chaotropic conditions, eliminating the use of toxic organic compounds or solvents. Mitochondrial DNA purified by this kit is suitable for various downstream molecular biology applications such as PCR, cloning, DNA hybridization, Southern Blotting, sequencing, genotyping, enzymatic analysis, and more.
DNA, no genomic DNA contamination
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty