Would you like to log in again?
ATPase Activity Assay Kit (Colorimetric), Biovision, Inc.
Supplier: BioVision
AB234055-100TEST
76186-196EA 648.23 USD InStock
76186-196
ATPase Activity Assay Kit (Colorimetric), Biovision, Inc.
Assays Enzyme Assays
Quick and easy method for monitoring ATPase activity in various samples.
  • Simple procedure; takes less than 1 hour
  • Fast and convenient
ATPase (Adenosine Triphosphatase: EC 3.6.1.3) is an important enzyme for maintaining the cell membrane potential, transporting ions and regulating cellular volume. It catalyzes the decomposition of ATP into ADP and a free phosphate ion. The hydrolysis of ATP is highly exergonic releasing energy that is utilized in several cellular processes. There are many classes of ATPases including Na+/K+ -ATPase, H+/K+ -ATPase, Ca2+ -ATPase, etc. The deficiency of mitochondrial ATPase is serious: for example, Na+/K+-ATPase deficiency increases anxiety-related behavior, while Ca2+ -ATPase deficiency leads to exertional muscle pain syndrome. Therefore, accurate detection of ATPase activity is valuable for the diagnosis and mechanistic studies of some of these diseases. BioVision’s ATPase Activity Assay kit provides a quick and easy method for monitoring ATPase activity in various samples. In the assay, ATPase hydrolyzes ATP releasing ADP and a free phosphate ion, and through linked reactions, a strong, stable chromophore is generated (OD 650 nm). The assay is simple, sensitive, high -throughput adaptable and can detect ATPase Activity less than 0.005 U/L
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty