Would you like to log in again?
Glyoxalase II Activity Kit (Colorimetric), Biovision, Inc.
Supplier: BioVision
AB273319-100TEST
76186-186EA 963.8 USD InStock
76186-186
Glyoxalase II Activity Kit (Colorimetric), Biovision, Inc.
Assays Enzyme Assays
The most sensitive kit in the market to detect Glyoxalase II activity
  • Simple procedure; takes ~ 40 minutes
  • Fast and convenient
  • Kit contains all necessary reagents for accurate measurement of Glyoxalase II
Glyoxalase II (GloII, hydroxyacylglutathione hydrolase, EC 3.1.2.6) is one of the two enzymes in the Glyoxalase system that is ubiquitously expressed in mammalian, plants and bacteria. The Glyoxalase system is a biological pathway that detoxifies cells by metabolizing α-ketoaldehydes such as methylglyoxal, a cytotoxic byproduct of lipids and glucose metabolism. GloII fulfills the terminal step in the glyoxalase system: It catalyzes the hydrolysis of S-D-lactoyl-glutathione (SLG) to reduced glutathione and D-lactic acid. The alteration of GloII activity in cell growth cycle, cell differentiation, phagocyte activation and diabetes mellitus development of implies this enzyme plays a wide-ranging and important role in cell function and disease progression. Research areas of current interest in Glo-II include diabetes, and cancer therapy. Biovision’s Glyoxalase II Activity Kit utilizes the ability of an active Glo-II to cleave a substrate while producing D-lactate. The produced D-Lactate reacts with the chromophore generating a stable signal that can be easily quantified at 450 nm using a microplate reader. Our assay kit is simple, sensitive and can detect as low as 0.6 mU/ml of Glo-II activity in biological samples
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty