Would you like to log in again?
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric), Biovision, Inc.
Supplier: BioVision
AB284550-100TEST
76186-202EA 819.5 USD InStock
76186-202
Carbonic Anhydrase (CA) Inhibitor Screening Kit (Colorimetric), Biovision, Inc.
Assays Enzyme Assays
High-throughput compatible and easy screening for Carbonic Anhydrase activity in biological samples
  • Simple, highly sensitive, high-throughput compatible
  • Easy protocol for the detection and quantification of active CA
  • Kit includes Acetazolamide, a CA inhibitor, and a stable, recombinant human CA
Carbonic anhydrases (CA) (CA, 4.2.1.1) are zinc enzymes present in both prokaryotes and eukaryotes. They catalyze efficiently the reversible hydration of CO2 to bicarbonate. Their important patho-physiological roles in respiration, pH and CO2 homeostasis, secretion, gluconeogenesis, ureagenesis etc. make it an important drug target. Characteristic increase and decrease of CA Activity is observed in different physiological conditions and diseases such asanemia, Thalassemia, Hypothyroidism, Hyperthyroidism, in several cases of lung and liver diseases and Leukemia. BioVision’s CA Activity Assay Kit can be used to measure CA activity in biological samples like serum and hemolysates. The assay utilizes the esterase activity of an active CA on an ester substrate which releases a chromophore. The released product can be easily quantified using an absorbance microplate reader. In the presence of a CA specific inhibitor, the enzyme loses its activity which results in a decrease of absorbance. This assay kit is simple and can be used to measure CA activity in biological samples like hemolysate, serum and purified CA activity, CA activity in biological samples like hemolysate and serum in a high-throughput format. To measure the Specific CA activity we have used the specific inhibitor Acetazolamide, which is a potent inhibitor of CA I, CA II, CA IV, CA IX, CA XII etc. except CA III
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty