Would you like to log in again?
Human Lactate Dehydrogenase A Inhibitor Screening Kit (Colorimetric), Biovision, Inc.
Supplier: BioVision
AB283393-100TEST
76186-158EA 997.72 USD InStock
76186-158
Human Lactate Dehydrogenase A Inhibitor Screening Kit (Colorimetric), Biovision, Inc.
Assays Enzyme Assays
Industry's first kit to screen/study/characterize potential inhibitors of LDH
  • Simple and sensitive assay.
  • Fast protocol - can be performed in 30 min.
  • High-throughput compatible
The human oxidoreductase Lactate Dehydrogenase A (LDHA) (EC:1.1.1.27) is a key enzyme in lactic acid metabolism. It interconverts pyruvate and NADH to lactate and NAD+ respectively. In mammals, LDHA exists as a tetramer and is primarily found in skeletal muscle tissues. LDHA is a common marker to assay for tissue damage and is also known to promote tumor proliferation in breast, lung, and pancreatic cancer patients. In BioVision’s LDHA Inhibitor Screening Kit, LDHA catalyzes the conversion of lactate to an intermediate that interacts with a probe to develop a color that can be measured at OD 450 nm. In the presence of a LDHA specific inhibitor, the reaction is impeded. A LDHA Inhibitor Control is included to compare the efficacy of the sample inhibitors. The assay is high-throughput adaptable and can be completed in less than 1 hr
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty