Would you like to log in again?
Oxalate Oxidase Activity Assay Kit (Fluorometric), Biovision
Supplier: BioVision
AB241007-100TEST
76186-220EA 798.73 USD InStock
76186-220
Oxalate Oxidase Activity Assay Kit (Fluorometric), Biovision
Assays Enzyme Assays
Simple, sensitive and can detect less than 4 µU of oxalate Oxidase activity per sample.
  • Rapid & Convenient
  • The assay can detect as little as 4 µU of OxOx enzymatic activity.
  • Includes Active OxOx Enzyme as Positive Control
Oxalate Oxidase (OxOx EC 1.2.3.4) belongs to the cupin protein superfamily and catalyzes the conversion of Oxalate into H2O2 and CO2. It plays an important role for stress response in plants. For example, wood rotting fungi generates high levels of oxalate, causing plant rot in many crops including lettuce, soybean, dry bean, tomato, etc. Overexpression of OxOx in plants (e.g. tomato, soybean, lettuce and tobacco) is able to generate transgenic plants showing resistance to fungal pathogenesis. Accurate measurement of Oxalate Oxidase activity is valuable for mechanistic studies in plants and development of transgenic plants. BioVision’s Oxalate Oxidase Activity Assay kit provides a quick and easy method for the measurement of Oxalate Oxidase activity in various samples. In this assay, Oxalate Oxidase converts oxalate into hydrogen peroxide, which in turn, reacts with a probe and converter generating a fluorometric signal (535/587 nm). The generated fluorescence is directly proportional to the amount of active Oxalate Oxidase present in samples. The assay is simple, sensitive, high –throughput adaptable and can detect less than 4 µU of oxalate Oxidase activity per sample.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty