Would you like to log in again?
PicoProbe™ Malate Dehydrogenase Activity Assay Kit (Fluorometric), Biovision, Inc.
Supplier: BioVision
AB235593-100TEST
76186-156EA 1034.21 USD InStock
76186-156
PicoProbe™ Malate Dehydrogenase Activity Assay Kit (Fluorometric), Biovision, Inc.
Assays Enzyme Assays
Simple, sensitive and high-throughput adaptable and can detect as low as 40 µU of MDH activity.
  • Simple, rapid & convenient
  • The assay can detect Malate Dehydrogenase (MDH) Activity less than 100 µU in various sample types.
Malate Dehydrogenase (MDH) (EC 1.1.1.37) reversibly converts L-Malate into oxaloacetate in the presence of NAD+. In eukaryotic cells, malate dehydrogenase has 2 isoforms, MDH1 and MDH2. MDH1 is cytosolic and participates in the malate-aspartate shuttle, which transports malate into the mitochondria. Malate will then be utilized for the generation of ATP. MDH2, on the other hand, is a mitochondrial enzyme that participates in the citric acid cycle. MDH2 activity is increased in some neurodegenerative diseases such as Alzheimer’s disease, and abnormal MDH activity in serum can serve as a diagnostic tool for severe liver damage (e.g. Hepatocellular carcinoma). Biovision’s PicoProbeTM Malate Dehydrogenase Activity Assay Kit (Fluorometric) provides a quick and easy method for monitoring MDH activities in a wide variety of samples. In this assay, MDH converts malate into intermediate products, which will further react with PicoProbeTM forming a fluorophore (Ex/Em=535/587nm). The kit is simple, sensitive and high-throughput adaptable and can detect as low as 40 µU of MDH activity.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty