Would you like to log in again?
Protein Carbonyl Content Assay Kit (Fluorometric), Biovision
Supplier: BioVision
AB235631-100TEST
76186-190EA 736.63 USD InStock
76186-190
Protein Carbonyl Content Assay Kit (Fluorometric), Biovision
Assays Protein Assays
The only, most sensitive kit in the market
  • Simple procedure; validated with human serum samples containing pmol of carbonylated protein
Protein carbonylation occurs when chronically elevated blood sugar levels create a microenvironment for sugars to covalently react with amino acids forming non-enzymatic adducts. Oxidative stress, the excessive production of ROS, can also give rise to the production of protein carbonyls. Over time, these two processess create Advanced Glycation End-products (AGEs) with proteins. The major functional outcome of protein-reactive AGEs is reduced enzymatic activity. Soluble AGEs can also bind and activate the receptor of advanced glycation end-products (RAGE). Receptor-dependent outcomes lead to tissue dysfunction, oxidative stress, and activation of monocytes, endothelial cells and mesangial cells. In Alzheimer’s Disease (AD) protein carbonyls are a marker of oxidative stress in the brain and are elevated in serum and plasma proportional to the cognitive severity. BioVision’s Protein Carbonyl Content Assay Kit uses Fluoroscein-5-Thiosemicarbazide (FTC), a fluorescent probe which covalently reacts with oxidized residues (i.e Cysteine, Lysine, Arginine, Histidine and Aspartic Acid) on proteins. The protein carbonyl content is determined by the generation of a stable fluorometric signal (Ex/Em 485/535 nm) and compared to the protein concentration determined in the BCA Assay to quantitate nmoles of carbonyl/mg protein. The kit is simple, requires no harsh chemicals, can quantitate carbonyls in serum or plasma and produces more reliable and reproducible results than the comparable colorimetric assays. It can detect carbonyl groups in samples with protein concentrations as low as 1 mg/ml!
Validated with human serum
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty