Would you like to log in again?
Glyoxalase I Activity Assay Kit (Colorimetric), Biovision, Inc.
Supplier: BioVision
AB241019-100TEST
76186-150EA 859.64 USD InStock
76186-150
Glyoxalase I Activity Assay Kit (Colorimetric), Biovision, Inc.
Assays Enzyme Assays
The most sensitive kit in the market to detect Glyoxalase I activity
  • Simple procedure; takes ~ 40 minutes
  • Fast and convenient
  • Kit contains all necessary reagents for accurate measurement of Glyoxalase I
Glyoxalase I (GloI, lactoylglutathione lyase, EC 4.4.1.5) is a cytosolic enzyme that participates in the Glyoxalase system. The glyoxalase system is a metabolic pathway that detoxifies α-ketoaldehydes such as methylglyoxal to D-lactic acid, via the intermediate S-D-lactoyl-glutathione (SLG). Specifically, GloI catalyzes the formation of SLG from methylglyoxal and reduced Glutathione. It is overexpressed in several human cancers such as colon, breast, prostate and melanoma. GloI also enhances resistance to antitumor agent-induced apoptosis in human leukemia cells. In addition, recent findings have linked GloI to numerous anxiety-like behaviors including psychiatric diseases (depression, schizophrenia and autism) and pain. BioVision’s Glyoxalase I Activity Assay Kit utilizes the ability of an active GloI to catalyze the formation of SLG by using two GloI substrates. The formation of the SLG can be tracked using a microplate reader at OD 240 nm. Our assay kit is simple, sensitive and can detect as low as 0.05 mU of Glo I activity in biological samples.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty