Would you like to log in again?
DL-Serine Assay Kit (Fluorometric), Biovision
Supplier: BioVision
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
AB241027-100TEST
76186-188EA 1155.01 USD InStock
76186-188
DL-Serine Assay Kit (Fluorometric), Biovision
Assays Cellular Assays
Market's first and only kit for enzymatic detection of Serine in biological samples
  • Simple, rapid & convenient assay
  • Limit of detection: 1 µM
  • Very specific, does not interact with other standard and non-standard amino acids
Serine is one of the 20 naturally-occurring amino acids used by all organisms in the biosynthesis of proteins. Having a single chiral center, serine can exist as either of two stereoisomers (L-Serine and D-Serine). In addition to its role as a protein building block, L-Serine serves as a precursor to many vital biomolecules, including phosphatidylserine, sphingomyelin and the amino acids glycine and cysteine. The non-proteogenic isomer D-Serine is also synthesized from L-Serine in the mammalian brain. While most D-amino acids are only present in trace amounts in mammals, D-Serine is a vital neurotransmitter that acts as a co-agonist of synaptic N-methyl-D-aspartate (NMDA) type glutamate-gated ion channels. Impaired D-Serine metabolism may contribute to several neuropsychiatric disorders, including depression, schizophrenia, epilepsy and dementia. BioVision’s DL-Serine Assay Kit allows for quantification of both L- and D-Serine in biological fluids and tissues. The assay is based on the conversion of L-Serine to D-Serine, which is metabolized to an intermediate product that is subsequently oxidized and reacts with a probe to form a stable fluorophore (Ex/Em = 539/587 nm). Samples may be divided and assayed simultaneously for quantification of both D-Serine and total DL-Serine. The assay is not affected by physiological concentrations of other amino acids, is high-throughput adaptable and can detect less than 1 μM Serine
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty