Would you like to log in again?
EZClick™ EdU Cell Proliferation/DNA Synthesis Kit (FACS/Microscopy), Red Fluorescence, Biovision, Inc.
Supplier: BioVision
AB287859-100TEST
76186-216EA 907.17 USD InStock
76186-216
EZClick™ EdU Cell Proliferation/DNA Synthesis Kit (FACS/Microscopy), Red Fluorescence, Biovision, Inc.
Assays Cellular Assays
Simple, fast, does not require lengthy incubation times.
All living cells undergo division cycle, a biological process crucial for proliferation and inheritance. Cell-division cycle is a series of events resulting in two daughter cells containing replicas of DNA from the original DNA molecule. DNA replication occurs in the S phase of the cell cycle and involves de novo synthesis of genomic DNA from its precursors. The ability of monitoring detailed characterization of cell cycle and DNA synthesis in proliferating cells is fundamental in basic, and applied immunologic and oncologic studies. Accurate determination of the effect of biologically active reagents on DNA synthesis and cell cycle is of great importance in anti-cancer drug discovery and basic biology. Biovision’s EZClick™ EdU DNA Synthesis Monitoring Kit utilizes a novel approach that relies on incorporation of 5-EdU (5-ethynyl-2’deoxyuridine) as nucleoside analog to thymidine into newly synthesized DNA directly in the cell culture. Incorporation of EdU into genomic DNA in S-phase is detected based on a click reaction between the alkyne moiety of EdU and fluorescent azide. Compared to historically used BrdU, click reaction is carried in mild conditions and flow cytometry/fluorescence microscopy can be used for assessment of proliferating cells in the population. Our kit provides sufficient materials for 100 assays based on the protocol.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty