Would you like to log in again?
Outdoor Round Vinyl Extension Cord, Woods Wire
Supplier: Woods Wire
860-529
300087-733EA 102.26 USD InStock
300087-733
Outdoor Round Vinyl Extension Cord, Woods Wire
Power Supplies
Made with stranded and cabled copper conductors
  • Solid molded plugs
  • Single outlet
  • 12/3 AWG conductor size
  • SJTW cable marking
Resists deterioration from moisture and abrasion.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty