Would you like to log in again?
Proto® TorquePlus™ Metric Combination Wrench Set, 18 Pieces, ORS Nasco
Supplier: ORS Nasco
Torqueplus™
577-1200R-MASD
300023-553EA 691.35 USD InStock
300023-553
Proto® TorquePlus™ Metric Combination Wrench Set, 18 Pieces, ORS Nasco
Wrenches
12 point set.
  • Increased torque applied to fastener with fewer rounded edges
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove to prevent slippage and knuckle injury
Delivery information: Opening sizes included are 7.000, 8.000, 9.000, 10.000, 11.000, 12.000, 13.000, 14.000, 15.000, 16.000, 17.000, 18.000, 19.000, 20.000, 21.000, 22.000, 23.000, and 24.000 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty