Would you like to log in again?
Proto® Crowfoot Wrenches, ORS Nasco
Supplier: ORS Nasco
577-5376CF
300026-590EA 180.9 USD InStock
300026-590
Proto® Crowfoot Wrenches, ORS Nasco
Wrenches
Anti-Slip Design open end (ASD) has patented locking grooves that help prevent the wrench from slipping off the fastener when turning
  • Full polish finish provides corrosion resistance and easy-to-clean surfaces
  • 2.375" opening size
  • Forged alloy steel construction, nickel chrome finish
  • 0.5" square drive type
  • Measurements provided in inches
Crowfoot wrenches are used in hard to reach, restricted access situations where a standard wrench or ratchet cannot reach.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty