Would you like to log in again?
SCRUBS® Stainless Steel Cleaner Towels, Essendant
Supplier: Essendant
SCRUBS®
ITW91930CT
102866-290CS 103.32 USD InStock
102866-290
SCRUBS® Stainless Steel Cleaner Towels, Essendant
Wipes Disposable Wipes and Towels Presaturated Wipes
Stainless steel cleaner towels clean, polish, brighten and protect in one step without dripping or overspraying.
  • Convenient premoistened towels
  • Prevents water marks and corrosion
  • Preserves high luster of stainless steel
  • For use on stainless steel, chrome and aluminum
  • Wet wipes
  • Polypropylene material
Chemical compound: D-Limonene; Mineral Oil
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty