Would you like to log in again?
Whatman™ Grade 0858 ½ Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10334347 10334352 10334353 10334351 10334353 10334345 10334351 10334352 10334345 10334347
10034-748PK 135.41 USD InStock
CA11013-954 CA11013-896 10034-748 10035-644 CA11013-374 CA11013-384 10034-746 10035-640 CA11013-380 10035-642
Whatman™ Grade 0858 ½ Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
Grained, with medium fast flow rate and medium retention. A universal filter paper, used for the filtration of extracts, oils, beer, syrups etc. Also applied in filter presses or for the aspiration of liquids.
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself
  • The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles
Time-saving Whatman qualitative grades are offered in this convenient form, which gives major advantages over flat circles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty