Would you like to log in again?
Whatman™ Grade 591 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
10311387 10311387
11008-572PK 1379.26 USD InStock
11008-572 CA11008-572
Whatman™ Grade 591 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
These cellulose filters are used in qualitative analytical techniques to determine and identify materials.
  • Thick filter paper with very high loading capacity
  • High absorbency and increased wet strength
A standard grade filter for fast filtration of medium to coarse precipitates.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty