Would you like to log in again?
Axygen® Hamilton CO-RE Style Automation Tips, Corning
Supplier: Corning
Axygen®
HTF-1000-C-HTR-S
10025-052CS 1003.14 USD InStock
10025-052
Axygen® Hamilton CO-RE Style Automation Tips, Corning
Pipette Tips
Axygen® automation tips are engineered to the exact specifications of the original equipment manufacturers’ tips, ensuring complete compatibility and a consistent fit.
  • Engineered to the exact specifications
  • Complete protection against contamination
Axygen® offers an alternative choice for the Hamilton® STARline system. These tips are manufactured to stringent specifications under strict process controls and assembled via automation to ensure consistent performance and quality. These clear polypropylene tips are available in a 96-well hanging tip rack, designed for Hamilton® MICROLAB® STAR, STARlet, STARplus, and Nimbus® workstations.
Certifications: Tips are certified RNase-/DNase-free, and nonpyrogenic.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty