Would you like to log in again?
Sterile Carbon Steel Surgical Blades, Propper Manufacturing
Supplier: Propper
12201500 12202100 12202000 12201200 12201000 12201100
76044-636BX 98.64 USD InStock
76044-636 76044-632 76044-634 76044-682 76044-684 76044-630
Sterile Carbon Steel Surgical Blades, Propper Manufacturing
Scalpels and Scalpel Assemblies Scalpel Blades
Propper Sterile Carbon Steel Surgical Blades are available in 7 sizes for every need.
  • Superior cutting ability of carbon steel
  • Sterile blades
  • Enclosed in heat-sealed peel-pack
Description of use of each size
#10: Curved cutting edge is one of the more traditional blade shapes, making small incisions to the skin and muscle clean and precise.
#11: Elongated triangular blade with a strong tip, ideal for stab incisions. Most commonly used for the incisions of chest drains, and opening various coronary arteries.
#15: Small curved cutting edge is the most popular blade for short and precise incisions.
#20: A larger version of the No. 10 blade equipped with a curved cutting edge, used mostly in orth0pedic and general surgical procedures
#21: Similar to the No. 21 blade with a curved cutting edge as well as a flat unsharpened back. Used for making small incisions in the skin and muscle.
#22: A tad bigger than the number #21 blade, used for skin incisions in both cardiac and thoracic surgery.
#23: A "leaf shaped" blade sharpened along its leading edge. Used for making long incisions such as an upper midline incision of the abdomen.
#24: Semi-circular shape, sharpened along its leading edge. Used for making long incisions in general surgery and also in autopsy procedures.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty