Would you like to log in again?
Leather Impact Drivers with Kevlar® Liner, Wells Lamont®
Supplier: Wells Lamont
I2430L I2430M I2430XXL I2430XL I2430S
76196-510PK 743.1 USD InStock
76196-510 76196-508 76196-506 76196-504 76196-512
Leather Impact Drivers with Kevlar® Liner, Wells Lamont®
Gloves
Amazing impact resistance and comfort.
  • Goatskin leather for comfort & durability
  • Kevlar® liner for cut protection
  • Soft and flexible TPR pads for impact protection on back of hands and fingers
  • Gunn cut
  • Straight thumb
Impact resistant work gloves deliver an extra level of protection to the back of the hand and fingers
These gloves take the standard leather driver to the next level and modernize it to meet the demanding standards set by refineries and construction sites. Applications include use in metal handling, construction, and pipe fitting.
The I2430 is constructed of grain goatskin leather and offers comfort, flexibility, and durability. A Kevlar® liner boosts protection to ANSI cut level 3 and thermoplastic (TPR) pads provide superior impact protection.
Certifications: ANSI Cut Level A3.
Ordering information: Sold by the dozen.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty