Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydrazinopyrimidine 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydrazinylpyrimidine

AC45305-0010 AC45305-0050
103464-676EA 237 USD InStock
103464-676 103464-674
2-Hydrazinopyrimidine 98%
2-Hydrazinopyrimidine


Formula: C₄H₆N₄
MW: 110.12 g/mol
Melting Pt: 110…112 °C
Storage Temperature: Ambient
MDL Number: MFCD01249337
CAS Number: 7504-94-1
Order Now
Specification Test Results
Appearance White to off-white to light brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty