Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cefoxitin sodium 94%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: MefoxinCefoxitin sodium saltSodium (6R,7S)-3-((carbamoyloxy)methyl)-7-methoxy-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

AC45528-0010 AC45528-0050
103518-204EA 330 USD InStock
103518-204 103518-202
Cefoxitin sodium 94%
Cefoxitin sodium


Formula: C₁₆H₁₆N₃NaO₇S₂
MW: 449.428 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00079042
CAS Number: 33564-30-6
Order Now
Specification Test Results
Appearance White to cream Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥93 %
Specific optical rotation +206° to +214° (20 °C, 589 nm) (c= 1, methanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty