Would you like to log in again?
Centrifuge Tube, Polystyrene, Globe Scientific
Supplier: Globe Scientific
112009
10022-252CS 201.28 USD InStock
10022-252
Centrifuge Tube, Polystyrene, Globe Scientific
Tubes Centrifuge Tubes
The 12 ml general-purpose centrifuge tubes are offered in polystyrene (PS).
Tubes are graduated at 0.5, 1.0, 2.5, 5.0, and 10.0 ml. Tubes are supplied with a plug cap, packaged separately within the same case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty