Would you like to log in again?
Proto® Center Punches, ORS Nasco
Supplier: ORS Nasco
577-417/16 577-413/8 577-411/2 577-415/8 577-415/16 577-411/4
76190-688EA 13.63 USD InStock
76190-688 76188-822 76188-820 76188-818 76190-690 76188-816
Proto® Center Punches, ORS Nasco
Chisels
Modified parabolic striking end for greater resistance to mushrooming and chipping.
  • Center punch
  • Round tip
  • Hex stock shape
  • Tool steel
For metal working layout such as marking intersecting lines, hole locations, and pilot drill starting points.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty